3',4'-Difluoro-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

29560-1G 29560-25G 29560-500MG 29560-5G
76913-340EA 185.66 USD
76913-340 76913-342 76913-344 76913-346
3',4'-Difluoro-[1,1'-biphenyl]-4-ol ≥97%
3',4'-Difluoro-[1,1'-biphenyl]-4-ol
Formula: C₁₂H₈F₂O
MW: 206.188 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16484256
CAS Number: 108185-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR