4-(2,3-Difluorophenyl)phenol ≥97%
Supplier: AOBChem USA
|
Warning
|
29546-1G
29546-25G
29546-500MG
29546-5G
76913-320EA
234.46
USD
76913-320
76913-322
76913-324
76913-326
4-(2,3-Difluorophenyl)phenol ≥97%
4-(2,3-Difluorophenyl)phenol
Formula:
C₁₂H₈F₂O MW: 206.188 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18312828 CAS Number: 202464-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...