4-(Benzyloxy)-3,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-3,5-dimethylbenzaldehyde

29531-25G 29531-5G
76913-310EA 381.93 USD
76913-310 76913-312
4-(Benzyloxy)-3,5-dimethylbenzaldehyde ≥95%
4-(Benzyloxy)-3,5-dimethylbenzaldehyd

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800683
CAS Number: 144896-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR