3-Bromoquinolin-7-amine ≥97% research grade

Supplier: AOBChem USA

Warning

29381-1G 29381-25G 29381-500MG 29381-5G
76913-264EA 257.8 USD
76913-264 76913-266 76913-268 76913-270
3-Bromoquinolin-7-amine ≥97% research grade
3-Bromoquinolin-7-amine
Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD20327980
CAS Number: 1344046-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR