3,6-Difluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29248-1G 29248-500MG 29248-5G
76913-226EA 528.33 USD
76913-226 76913-228 76913-230
3,6-Difluoro-2-methylbenzoic acid ≥97%
3,6-Difluoro-2-methylbenzoic acid
Formula: C₈H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD23712074
CAS Number: 459836-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR