4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Trifluoromethylphenyl)benzaldehyde, 4'-Trifluoromethyl-biphenyl-4-carbaldehyde, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

29071-25G 29071-5G
76913-130EA 289.63 USD
76913-130 76913-132
4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥97%
4'-Trifluoromethylbiphenyl-4-carbaldehyde
Formula: C₁₄H₉F₃O
MW: 250.2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862519
CAS Number: 90035-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR