3-Cyclopentylphenylboronic acid pinacol ester ≥95% research grade

Supplier: AOBChem USA

Warning

29003-1G 29003-500MG 29003-5G
76913-106EA 772.34 USD
76913-106 76913-108 76913-110
3-Cyclopentylphenylboronic acid pinacol ester ≥95% research grade
3-Cyclopentylphenylboronic acid pinacol ester
Formula: C₁₇H₂₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD28806017
CAS Number: 2121514-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR