3'-Bromo-5'-chloroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Bromo-5-chlorophenyl)ethanone

28858-1G 28858-25G 28858-500MG 28858-5G
76913-070EA 138.98 USD
76913-070 76913-072 76913-074 76913-076
3'-Bromo-5'-chloroacetophenone ≥97%
3'-Bromo-5'-chloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189467
CAS Number: 154257-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR