3-Hydroxy-2-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

28714-1G 28714-500MG 28714-5G
76913-044EA 825.39 USD
76913-044 76913-046 76913-048
3-Hydroxy-2-methylphenylboronic acid ≥97% research grade
3-Hydroxy-2-methylphenylboronic acid
Formula: C₇H₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18397306
CAS Number: 948592-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR