3-Hydroxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28702-1G 28702-25G 28702-5G
76913-034EA 96.55 USD
76913-034 76913-036 76913-038
3-Hydroxy-5-methylbenzaldehyde ≥97%
3-Hydroxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999707
CAS Number: 60549-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR