4-(4,4-Dimethylcyclohexyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

28620-1G 28620-500MG 28620-5G
76913-026EA 528.33 USD
76913-026 76913-028 76913-030
4-(4,4-Dimethylcyclohexyl)phenylboronic acid ≥95%
4-(4,4-Dimethylcyclohexyl)phenylboronic acid
Formula: C₁₄H₂₁BO₂
MW: 232.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30529801
CAS Number: 865360-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR