3-Bromo-5-iodopyridine-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

28419-1G 28419-500MG 28419-5G
76913-002EA 683.22 USD
76913-002 76913-004 76913-006
3-Bromo-5-iodopyridine-4-carboxylic acid ≥95%
3-Bromo-5-iodopyridine-4-carboxylic acid
Formula: C₆H₃BrINO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28360689
CAS Number: 1809168-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR