3-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-fluorophenylboronic acid pinacol ester, 2-(3-Chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

28237-1G 28237-25G 28237-5G
76912-976EA 74.27 USD
76912-976 76912-978 76912-980
3-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥97%
3-Chloro-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClFO₂
MW: 256.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405368
CAS Number: 1192025-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR