3-Fluoro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

28189-1G 28189-25G 28189-500MG 28189-5G
76912-964EA 231.28 USD
76912-964 76912-966 76912-968 76912-970
3-Fluoro-2-iodophenol ≥97%
3-Fluoro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
Storage Temperature: Refrigerator
MDL Number: MFCD08166555
CAS Number: 863870-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR