3-Bromo-2-iodobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

28113-1G 28113-500MG 28113-5G
76912-950EA 528.33 USD
76912-950 76912-952 76912-954
3-Bromo-2-iodobenzonitrile ≥97%
3-Bromo-2-iodobenzonitrile
Formula: C₇H₃BrIN
Storage Temperature: Ambient
MDL Number: MFCD18391644
CAS Number: 450412-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR