3-Bromo-2,6-difluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

28037-25G 28037-5G
76912-938EA 527.27 USD
76912-938 76912-940
3-Bromo-2,6-difluorobenzonitrile ≥97%
3-Bromo-2,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MW: 217.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987589
CAS Number: 1250444-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR