4-Bromo-2,6-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27936-25G 27936-5G
76912-918EA 371.32 USD
76912-918 76912-920
4-Bromo-2,6-dimethylbenzoic acid ≥97%
4-Bromo-2,6-dimethylbenzoic acid
Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09965997
CAS Number: 74346-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR