3-Fluoro-5-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

27872-1G 27872-500MG 27872-5G
76912-908EA 558.04 USD
76912-908 76912-910 76912-912
3-Fluoro-5-(methylthio)phenol ≥97%
3-Fluoro-5-(methylthio)phenol
Formula: C₇H₇FOS
Storage Temperature: Refrigerator
MDL Number: MFCD16998929
CAS Number: 1243456-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR