2-Iodo-4-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Trifluoromethoxy)-2-iodoaniline

27715-25G 27715-5G
76912-854EA 262.05 USD
76912-854 76912-856
2-Iodo-4-(trifluoromethoxy)aniline ≥97%
2-Iodo-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃INO
Storage Temperature: Ambient
MDL Number: MFCD04972994
CAS Number: 845866-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR