3-Fluoro-4-(trimethylsilyl)pyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

27495-1G 27495-25G 27495-500MG 27495-5G
76912-832EA 379.81 USD
76912-832 76912-834 76912-836 76912-838
3-Fluoro-4-(trimethylsilyl)pyridine-2-carboxylic acid ≥97%
3-Fluoro-4-(trimethylsilyl)pyridine-2-carboxylic acid
Formula: C₉H₁₂FNO₂Si
MW: 213.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142595
CAS Number: 1809168-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR