3-(N,N-Dimethylaminomethyl)-2-fluorophenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
27368-1G
27368-25G
27368-500MG
27368-5G
76912-794EA
207.94
USD
76912-794
76912-796
76912-798
76912-800
3-(N,N-Dimethylaminomethyl)-2-fluorophenylboronic acid ≥95%
3-(N,N-Dimethylaminomethyl)-2-fluorophenylboronic acid
Formula:
C₉H₁₃BFNO₂ MW: 197.014 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28142592 CAS Number: 2121513-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...