3-Bromo-6-fluoro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

27303-1G 27303-25G 27303-5G
76912-764EA 64.72 USD
76912-764 76912-766 76912-768
3-Bromo-6-fluoro-2-methoxyphenol ≥97%
3-Bromo-6-fluoro-2-methoxyphenol
Formula: C₇H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12910488
CAS Number: 1026796-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR