3-Chloro-4-ethoxy-phenol ≥97%

Supplier: AOBChem USA

Warning

27297-1G 27297-25G 27297-500MG 27297-5G
76912-748EA 162.32 USD
76912-748 76912-750 76912-752 76912-754
3-Chloro-4-ethoxy-phenol ≥97%
3-Chloro-4-ethoxy-phenol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14536966
CAS Number: 1216134-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR