3-Ethoxy-5-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

27271-1G 27271-25G 27271-500MG 27271-5G
76912-724EA 198.39 USD
76912-724 76912-726 76912-728 76912-730
3-Ethoxy-5-fluorobenzaldehyde ≥95%
3-Ethoxy-5-fluorobenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375377
CAS Number: 1378670-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR