4-Bromo-2-chloro-3-iodophenol ≥97%

Supplier: AOBChem USA

Warning

27253-1G 27253-25G 27253-500MG 27253-5G
76912-710EA 171.87 USD
76912-710 76912-712 76912-714 76912-716
4-Bromo-2-chloro-3-iodophenol ≥97%
4-Bromo-2-chloro-3-iodophenol
Formula: C₆H₃BrClIO
Storage Temperature: Refrigerator
MDL Number: MFCD28015444
CAS Number: 1809158-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR