3,5-Dichloro-4-methoxybenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

27233-1G 27233-25G 27233-500MG 27233-5G
76912-696EA 171.87 USD
76912-696 76912-698 76912-700 76912-702
3,5-Dichloro-4-methoxybenzyl alcohol ≥95%
3,5-Dichloro-4-methoxybenzyl alcohol
Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03997279
CAS Number: 4892-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR