3,4,5-Trichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27209-25G 27209-5G
76912-688EA 481.65 USD
76912-688 76912-690
3,4,5-Trichlorobenzaldehyde ≥97%
3,4,5-Trichlorobenzaldehyde
Formula: C₇H₃Cl₃O
MW: 209.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18400105
CAS Number: 56961-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR