3-Bromo-5-fluoro-N-methylaniline ≥97%

Supplier: AOBChem USA

Warning

27082-1G 27082-25G 27082-500MG 27082-5G
76912-650EA 292.81 USD
76912-650 76912-652 76912-654 76912-656
3-Bromo-5-fluoro-N-methylaniline ≥97%
3-Bromo-5-fluoro-N-methylaniline
Formula: C₇H₇BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD09999730
CAS Number: 1369880-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR