3-Fluoro-5-(pyrrolidino)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

27079-1G 27079-25G 27079-500MG 27079-5G
76912-636EA 260.99 USD
76912-636 76912-638 76912-640 76912-642
3-Fluoro-5-(pyrrolidino)phenylboronic acid pinacol ester ≥97%
3-Fluoro-5-(pyrrolidino)phenylboronic acid pinacol ester
Formula: C₁₆H₂₃BFNO₂
MW: 291.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039611
CAS Number: 1415928-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR