4-Bromo-2,5-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27042-1G 27042-25G 27042-500MG 27042-5G
76912-622EA 155.96 USD
76912-622 76912-624 76912-626 76912-628
4-Bromo-2,5-dimethoxybenzoic acid ≥97%
4-Bromo-2,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779240
CAS Number: 35458-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR