4-Bromo-2-chloro-5-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

26978-1G 26978-25G 26978-5G
76912-602EA 96.55 USD
76912-602 76912-604 76912-606
4-Bromo-2-chloro-5-fluoropyridine ≥97%
4-Bromo-2-chloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MW: 210.431 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659500
CAS Number: 884495-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR