3-Methyl-5-(methylthio)benzaldehyde ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
26913-1G
26913-25G
26913-5G
76912-558EA
82.76
USD
76912-558
76912-560
76912-562
3-Methyl-5-(methylthio)benzaldehyde ≥97% research grade
3-Methyl-5-(methylthio)benzaldehyde
Formula:
C₉H₁₀OS Storage Temperature: Ambient |
MDL Number:
MFCD27756685 CAS Number: 1289089-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...