4-(Benzoyloxy)cyclohexanone ≥97%

Supplier: AOBChem USA

Warning

26753-1G 26753-25G 26753-500MG 26753-5G
76912-514EA 320.4 USD
76912-514 76912-516 76912-518 76912-520
4-(Benzoyloxy)cyclohexanone ≥97%
4-(Benzoyloxy)cyclohexanone
Formula: C₁₃H₁₄O₃
MW: 218.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00458687
CAS Number: 23510-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR