3-Amino-4-methylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
26557-1G
26557-25G
26557-5G
76912-446EA
84.88
USD
76912-446
76912-448
76912-450
3-Amino-4-methylphenylboronic acid pinacol ester ≥97%
3-Amino-4-methylphenylboronic acid pinacol ester
Formula:
C₁₃H₂₀BNO₂ MW: 233.114 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22493533 CAS Number: 850689-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...