4-[(3,4-Dichlorophenyl)methyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

26548-1G 26548-25G 26548-500MG 26548-5G
76912-434EA 242.95 USD
76912-434 76912-436 76912-438 76912-440
4-[(3,4-Dichlorophenyl)methyl]morpholine ≥97%
4-[(3,4-Dichlorophenyl)methyl]morpholine
Formula: C₁₁H₁₃Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD00979502
CAS Number: 212392-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR