3-Fornyl-4-morpholinophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26541-1G 26541-25G 26541-500MG 26541-5G
76912-420EA 320.4 USD
76912-420 76912-422 76912-424 76912-426
3-Fornyl-4-morpholinophenylboronic acid pinacol ester ≥97%
3-Fornyl-4-morpholinophenylboronic acid pinacol ester
Formula: C₁₇H₂₄BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD21333040
CAS Number: 2096330-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR