3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-cyanophenylboronic acid pinacol ester

26507-1G 26507-25G 26507-500MG 26507-5G
76912-402EA 157.02 USD
76912-402 76912-404 76912-406 76912-408
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formula: C₁₃H₁₅BClNO₂
MW: 263.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659095
CAS Number: 1212021-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR