3-Amino-2-bromo-6-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Amino-6-chloro-2-bromopyridine, 2-Bromo-6-chloro-3-pyridylamine, 2-Bromo-6-chloropyridin-3-amine

26472-25G 26472-5G
76912-392EA 310.85 USD
76912-392 76912-394
3-Amino-2-bromo-6-chloropyridine ≥97%
3-Amino-2-bromo-6-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504927
CAS Number: 1050501-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR