1-Bromo-3-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

26446-25G 26446-5G
76912-384EA 212.18 USD
76912-384 76912-386
1-Bromo-3-fluoro-5-iodobenzene ≥97%
1-Bromo-3-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
Storage Temperature: Ambient
MDL Number: MFCD06657969
CAS Number: 845866-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR