4-Bromo-2-(1-methylethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

26373-1G 26373-25G 26373-500MG 26373-5G
76912-360EA 429.67 USD
76912-360 76912-362 76912-364 76912-366
4-Bromo-2-(1-methylethyl)benzenemethanol ≥97%
4-Bromo-2-(1-methylethyl)benzenemethanol
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26045275
CAS Number: 1370600-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR