3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Amino-4-methylphenylboronic acid, pinacol ester, HCl

26355-1G 26355-25G 26355-5G
76912-346EA 111.4 USD
76912-346 76912-348 76912-350
3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride ≥97%
3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₁BClNO₂
MW: 269.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659921
CAS Number: 850567-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR