3-Bromo-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

26329-1G 26329-25G 26329-5G
76912-318EA 106.09 USD
76912-318 76912-320 76912-322
3-Bromo-2-fluorobenzamide ≥97%
3-Bromo-2-fluorobenzamide
Formula: C₇H₅BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD12913463
CAS Number: 871353-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR