3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzeneboronic acid, pinacol ester

26316-5G 26316-25G
76900-346EA 125.19 USD
76900-346 76912-308
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥97% research grade
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula: C₂₂H₃₅BN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16294542
CAS Number: 883738-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR