3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26291-1G 26291-25G 26291-500MG 26291-5G
76912-292EA 290.69 USD
76912-292 76912-294 76912-296 76912-298
3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester ≥97%
3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18732618
CAS Number: 1051316-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR