3-Hydroxy-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

26224-1G 26224-25G 26224-500MG 26224-5G
76912-268EA 320.4 USD
76912-268 76912-270 76912-272 76912-274
3-Hydroxy-5-phenylpyridine ≥97%
3-Hydroxy-5-phenylpyridine
Formula: C₁₁H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD04114220
CAS Number: 31676-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR