3,5-Bis(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

26212-1G 26212-25G 26212-500MG 26212-5G
76912-238EA 177.18 USD
76912-238 76912-240 76912-242 76912-244
3,5-Bis(methylthio)pyridine ≥95%
3,5-Bis(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NS₂
MW: 171.283 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015800
CAS Number: 70999-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR