4-Bromo-1-ethyl-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

26150-1G 26150-25G 26150-5G
76912-204EA 84.88 USD
76912-204 76912-206 76912-208
4-Bromo-1-ethyl-2-nitrobenzene ≥97%
4-Bromo-1-ethyl-2-nitrobenzene
Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490699
CAS Number: 10342-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR