3-Chloro-4-(N,N-dimethylamino)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26119-1G 26119-25G 26119-500MG 26119-5G
76912-192EA 350.1 USD
76912-192 76912-194 76912-196 76912-198
3-Chloro-4-(N,N-dimethylamino)phenylboronic acid pinacol ester ≥97%
3-Chloro-4-(N,N-dimethylamino)phenylboronic acid pinacol ester
Formula: C₁₄H₂₁BClNO₂
MW: 281.586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045257
CAS Number: 1613259-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR