4-Amino-3-isopropylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26103-1G 26103-25G 26103-500MG 26103-5G
76912-178EA 260.99 USD
76912-178 76912-180 76912-182 76912-184
4-Amino-3-isopropylphenylboronic acid pinacol ester ≥97%
4-Amino-3-isopropylphenylboronic acid pinacol ester
Formula: C₁₅H₂₄BNO₂
MW: 261.168 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26045250
CAS Number: 2121513-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR