3,5-Dibromo-4-cyanopyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
26088-25G
26088-5G
76912-156EA
244.01
USD
76912-156
76912-158
3,5-Dibromo-4-cyanopyridine ≥97%
3,5-Dibromo-4-cyanopyridine
Formula:
C₆H₂Br₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD09263321 CAS Number: 870244-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...